4-Bromo-1,3-dimethyl-1H-pyrazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HI-1487-5G HI-1487-1G
COBBHI-1487-5GEA 0 GBP
COBBHI-1487-5G COBBHI-1487-1G
4-Bromo-1,3-dimethyl-1H-pyrazole
4-Bromo-1,3-dimethyl-1H-pyrazole
Formula: C₅H₇BrN₂
MW: 175.0265 g/mol
MDL Number: MFCD00985488
CAS Number: 5775-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR