4-Bromo-1-methyl-1H-pyrazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HI-1308-5G HI-1308-25G
COBBHI-1308-5GEA 0 GBP
COBBHI-1308-5G COBBHI-1308-25G
4-Bromo-1-methyl-1H-pyrazole
4-Bromo-1-methyl-1H-pyrazole
Formula: C₄H₅BrN₂
MW: 161.00 g/mol
MDL Number: MFCD02179565
CAS Number: 15803-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR