4-Bromo-1-ethyl-1H-pyrazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromo-1-ethylpyrazole

HC-6396-25G HC-6396-1G HC-6396-5G
COBBHC-6396-25GEA 0 GBP
COBBHC-6396-25G COBBHC-6396-1G COBBHC-6396-5G
4-Bromo-1-ethyl-1H-pyrazole
4-Bromo-1-ethyl-1H-pyrazole
Formula: C₅H₇BrN₂
MW: 175.0265 g/mol
MDL Number: MFCD08235252
CAS Number: 71229-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR