2,3-Dimethylmaleic anhydride

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22821G100 AS22821G25
ACTIAS22821G100EA 0 GBP
ACTIAS22821G100 ACTIAS22821G25
2,3-Dimethylmaleic anhydride
2,3-Dimethylmaleic anhydride
Formula: C₆H₆O₃
MDL Number: MFCD00005523
CAS Number: 766-39-2
EINECS: 212-165-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR