3'-Fluoro-4'-(1-imidazolyl)acetophenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21557G25 AS21557G5
ACTIAS21557G25EA 0 GBP
ACTIAS21557G25 ACTIAS21557G5
3'-Fluoro-4'-(1-imidazolyl)acetophenone
3'-Fluoro-4'-(1-imidazolyl)acetophenone
Formula: C₁₁H₉FN₂O
MDL Number: MFCD11136857
CAS Number: 870838-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR