3'-Bromo-4'-(1-imidazolyl)acetophenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21560G1 AS21560G5
ACTIAS21560G1EA 0 GBP
ACTIAS21560G1 ACTIAS21560G5
3'-Bromo-4'-(1-imidazolyl)acetophenone
3'-Bromo-4'-(1-imidazolyl)acetophenone
Formula: C₁₁H₉BrN₂O
MDL Number: MFCD11656809
CAS Number: 1141669-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR