3,4-(Methylenedioxy)phenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,4-(Methylenedioxy)benzeneboronic acid, Benzo[d][1,3]dioxol-5-ylboronic acid

011004-1G 011004-5G 011004-25G
FLUO011004-1GEA 0 GBP
FLUO011004-1G FLUO011004-5G FLUO011004-25G
3,4-(Methylenedioxy)phenylboronic acid
3,4-(Methylenedioxy)phenylboronic acid
Formula: C₇H₇BO₄
MW: 165.94 g/mol
MDL Number: MFCD01009695
CAS Number: 94839-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR