1-(Bromophenyl)-4-piperidone

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AM-1717-5G AM-1717-1G AM-1717-250M
COBBAM-1717-5GEA 0 GBP
COBBAM-1717-5G COBBAM-1717-1G COBBAM-1717-250M
1-(Bromophenyl)-4-piperidone
1-(Bromophenyl)-4-piperidone
Formula: C₁₁H₁₂BrNO
MDL Number: MFCD09930458
CAS Number: 154913-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR