1H,1H,2H,2H-Perfluorooctyltriethoxysilane

Supplier: ABCR
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Triethoxy-1H,1H,2H,2H-tridecafluoro-n-octylsilane, Tridecafluoro-1,1,2,2-tetrahydrooctyl-1-triethoxysilane, Triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane;

AB104055.0100 AB104055.0005
ABCRAB104055.0100EA 0 GBP
ABCRAB104055.0100 ABCRAB104055.0005
1H,1H,2H,2H-Perfluorooctyltriethoxysilane
1H,1H,2H,2H-Perfluorooctyltriethoxysilane
Formula: C₁₄H₁₉F₁₃O₃Si
MW: 510.367 g/mol
MDL Number: MFCD00042333
CAS Number: 51851-37-7
EINECS: 257-473-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR