4-Amino-1-boc-piperidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20222G100 AS20222G25
ACTIAS20222G100EA 0 GBP
ACTIAS20222G100 ACTIAS20222G25
4-Amino-1-boc-piperidine
4-Amino-1-Boc-piperidine
Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD01076201
CAS Number: 87120-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR