(S)-(+)-3-Amino-1-boc-piperidine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
011309-1G
011309-5G
FLUO011309-1GEA
0
GBP
FLUO011309-1G
FLUO011309-5G
(S)-(+)-3-Amino-1-boc-piperidine
(S)-(+)-3-Amino-1-Boc-piperidine
Formula:
C₁₀H₂₀N₂O₂ |
MDL Number:
MFCD03094718 CAS Number: 625471-18-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...