4-(Boc-amino)piperidine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-N-(tert-Butoxycarbonyl)aminopiperidine, tert-Butyl piperidin-4-ylcarbamate, tert-butyl N-(piperidin-4-yl)carbamate, 4-Boc-Aminopiperidine, 4-N-BOC-Aminopiperidine
AS20200G100
AS20200G25
ACTIAS20200G100EA
93
GBP
ACTIAS20200G100
ACTIAS20200G25
4-(Boc-amino)piperidine
4-(Boc-amino)piperidine
Formula:
C₁₀H₂₀N₂O₂ MW: 200.28 g/mol |
MDL Number:
MFCD00798171 CAS Number: 73874-95-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...