(±)-3-(Boc-amino)piperidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: tert-Butyl piperidin-3-ylcarbamate, 3-Aminopiperidine, 3-BOC protected

AS21929G1 AS21929G25 AS21929G5
ACTIAS21929G1EA 0 GBP
ACTIAS21929G1 ACTIAS21929G25 ACTIAS21929G5
(±)-3-(Boc-amino)piperidine
(±)-3-(Boc-amino)piperidine
Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD03839941
CAS Number: 172603-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR