(S)-3-(Boc-amino)piperidine

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (S)-tert-Butyl piperidin-3-ylcarbamate(S)-3-Boc-aminopiperidinetert-butyl N-[(3S)-piperidin-3-yl]carbamate

ACTIAS12151G25EA 53 GBP
ACTIAS12151G1 ACTIAS12151G25 ACTIAS12151G5
(S)-3-(Boc-amino)piperidine
(S)-3-(Boc-amino)piperidine
Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
MDL Number: MFCD03093383
CAS Number: 216854-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR