N(α)-Cbz-L-histidine
Supplier: BACHEM BIOCHEMICA
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Cbz-His-OH, N(α)-Benzyloxycarbonyl-L-histidine, N-Cbz-L-histidine, Z-His-OH
4000625.0005
4000625.0025
BACE4000625.0005EA
0
GBP
BACE4000625.0005
BACE4000625.0025
N(α)-Cbz-L-histidine
N(α)-Cbz-L-histidine
Formula:
C₁₄H₁₅N₃O₄ MW: 289.3 g/mol |
MDL Number:
MFCD00065960 CAS Number: 14997-58-1 EINECS: 239-084-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...