Tin(IV) phthalocyanine dichloride

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

39606.03 39606.06
39606.03EA 67.7 GBP
39606.03 39606.06
Tin(IV) phthalocyanine dichloride
Tin (IV) phthalocyanine dichloride

CAS No.: 18253-54-8 Molecular Formula: C32H16Cl2N8Sn Formula Weight: 702.34 Physical Form: Powder Sensitivity: Moisture Sensitive MDL No.: MFCD00054210

Formula: C₃₂H₁₆Cl₂N₈Sn
MW: 702.139 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00054210
CAS Number: 18253-54-8
UN: 3146
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR