[1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

H26895.06 H26895.14
H26895.06EA 286 GBP
H26895.06 H26895.14
[1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride ≥99%
[1,1'-Bis(diphenylphosphino)ferrocene]nickel (II) chloride

Purity: 99% CAS No.: 67292-34-6 Molecular Formula: C34H28Cl2FeNiP2 Formula Weight: 684.00 Melting Point: 285° MDL No.: MFCD00270284

Formula: C₃₄H₂₈Cl₂FeNiP₂
Melting Pt: 285 °C
MDL Number: MFCD00270284
CAS Number: 67292-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR