Bis(2,2,2-trifluoroethyl)amine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004124-1G 004124-5G
FLUO004124-1GEA 0 GBP
FLUO004124-1G FLUO004124-5G
Bis(2,2,2-trifluoroethyl)amine
Bis(2,2,2-trifluoroethyl)amine
Formula: C₄H₅F₆N
MW: 181.08 g/mol
MDL Number: MFCD00042094
CAS Number: 407-01-2
EINECS: 206-978-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR