2'-Methoxy-5'-methyl-3'-nitroacetophenone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
019300-5G
019300-25G
FLUO019300-5GEA
0
GBP
FLUO019300-5G
FLUO019300-25G
2'-Methoxy-5'-methyl-3'-nitroacetophenone
2'-Methoxy-5'-methyl-3'-nitroacetophenone
Formula:
C₁₀H₁₁NO₄ |
MDL Number:
MFCD03789147 CAS Number: 103204-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...