3-Formyl-2-methoxyphenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-3486-25G
BB-3486-5G
BB-3486-1G
COBBBB-3486-25GEA
0
GBP
COBBBB-3486-25G
COBBBB-3486-5G
COBBBB-3486-1G
3-Formyl-2-methoxyphenylboronic acid
3-Formyl-2-methoxyphenylboronic acid
Formula:
C₈H₉BO₄ MW: 179.9700 g/mol |
MDL Number:
MFCD08689486 CAS Number: 480424-49-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...