3-Iodo-1H-pyrazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Iodopyrazole

HI-1600-5G HI-1600-1G
COBBHI-1600-5GEA 0 GBP
COBBHI-1600-5G COBBHI-1600-1G
3-Iodo-1H-pyrazole
3-Iodo-1H-pyrazole
Formula: C₃H₃IN₂
MW: 193.98 g/mol
MDL Number: MFCD12022324
CAS Number: 4522-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR