Cyclopentadienyltitanium trichloride
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
77134.03
77134.06
77134.03EA
39.6
GBP
77134.03
77134.06
Cyclopentadienyltitanium trichloride
Cyclopentadienyltitanium trichloride
CAS No.: 1270-98-0
Molecular Formula: C5H5Cl3Ti
Formula Weight: 219.35
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Appearance: Yellow-orange
Melting Point: 210° dec.
UN No.: 3261
MDL No.: MFCD00003724
Formula:
C₅H₅Cl₃Ti Melting Pt: 210 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003724 CAS Number: 1270-98-0 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...