4'-Hydroxyheptanophenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23120G100 AS23120G25
ACTIAS23120G100EA 0 GBP
ACTIAS23120G100 ACTIAS23120G25
4'-Hydroxyheptanophenone
4'-Hydroxyheptanophenone
Formula: C₁₃H₁₈O₂
MW: 206.2800 g/mol
MDL Number: MFCD00191523
CAS Number: 14392-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR