3'-Fluoro-2'-hydroxyacetophenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23039G1 AS23039G5
ACTIAS23039G1EA 0 GBP
ACTIAS23039G1 ACTIAS23039G5
3'-Fluoro-2'-hydroxyacetophenone
3'-Fluoro-2'-hydroxyacetophenone
Formula: C₈H₇FO₂
MW: 154.14 g/mol
MDL Number: MFCD09055140
CAS Number: 699-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR