2-Amino-4-(4-nitrophenyl)thiazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

015327-1G 015327-2.5G
FLUO015327-1GEA 0 GBP
FLUO015327-1G FLUO015327-2.5G
2-Amino-4-(4-nitrophenyl)thiazole
2-Amino-4-(4-nitrophenyl)thiazole
Formula: C₉H₇N₃O₂S
MDL Number: MFCD00170222
CAS Number: 2104-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR