Methyl-4-(bromomethyl)benzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21749G100 AS21749G25
ACTIAS21749G100EA 0 GBP
ACTIAS21749G100 ACTIAS21749G25
Methyl-4-(bromomethyl)benzoate
Methyl 4-(bromomethyl)benzoate
Formula: C₉H₉BrO₂
MDL Number: MFCD00032453
CAS Number: 2417-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR