4-Bromo-3-(trifluoromethyl)phenol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Bromo-5-hydroxybenzotrifluoride
AS25246G100
AS25246G25
AS25246G5
ACTIAS25246G100EA
290
GBP
ACTIAS25246G100
ACTIAS25246G25
ACTIAS25246G5
4-Bromo-3-(trifluoromethyl)phenol
4-Bromo-3-(trifluoromethyl)phenol
Formula:
C₇H₄BrF₃O MW: 241.01 g/mol |
MDL Number:
MFCD03095342 CAS Number: 320-49-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...