4-(Trifluoromethylthio)phenol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24179G100 AS24179G25
ACTIAS24179G100EA 0 GBP
ACTIAS24179G100 ACTIAS24179G25
4-(Trifluoromethylthio)phenol
4-(Trifluoromethylthio)phenol
Formula: C₇H₅F₃OS
MDL Number: MFCD00036035
CAS Number: 461-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR