1-Phenylcyclobutanecarboxylic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20006G1
AS20006G5
ACTIAS20006G1EA
83
GBP
ACTIAS20006G1
ACTIAS20006G5
1-Phenylcyclobutanecarboxylic acid
1-Phenylcyclobutanecarboxylic acid
Formula:
C₁₀H₁₀O₂ MW: 176.22 g/mol |
MDL Number:
MFCD00019262 CAS Number: 37828-19-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...