Benzofuran-2-carboxylic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21974G25 AS21974G5
ACTIAS21974G25EA 0 GBP
ACTIAS21974G25 ACTIAS21974G5
Benzofuran-2-carboxylic acid
Benzofuran-2-carboxylic acid
Formula: C₉H₆O₃
MDL Number: MFCD00005848
CAS Number: 496-41-3
EINECS: 207-818-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR