4-Aminopyrimidine-5-carbonitrile
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS23172G5
AS23172G25
ACTIAS23172G5EA
0
GBP
ACTIAS23172G5
ACTIAS23172G25
4-Aminopyrimidine-5-carbonitrile
4-Aminopyrimidine-5-carbonitrile
Formula:
C₅H₄N₄ MW: 120.11 g/mol |
MDL Number:
MFCD00054552 CAS Number: 16357-69-0 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...