4-Fluoro-D-phenylalanine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23100G25 AS23100G5
ACTIAS23100G25EA 0 GBP
ACTIAS23100G25 ACTIAS23100G5
4-Fluoro-D-phenylalanine
4-Fluoro-D-phenylalanine
Formula: C₉H₁₀FNO₂
MDL Number: MFCD00063063
CAS Number: 18125-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR