2-Methyl-5-nitrobenzotrifluoride
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS24116G25
AS24116G5
ACTIAS24116G25EA
0
GBP
ACTIAS24116G25
ACTIAS24116G5
2-Methyl-5-nitrobenzotrifluoride
2-Methyl-5-nitrobenzotrifluoride
Formula:
C₈H₆F₃NO₂ |
MDL Number:
MFCD01631684 CAS Number: 89976-12-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...