5-Amino-2-methylbenzotrifluoride

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24690G100 AS24690G25
ACTIAS24690G100EA 0 GBP
ACTIAS24690G100 ACTIAS24690G25
5-Amino-2-methylbenzotrifluoride
5-Amino-2-methylbenzotrifluoride
Formula: C₈H₈F₃N
MDL Number: MFCD01631582
CAS Number: 65934-74-9
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR