DL-tert-Leucine (H-DL-Tle-OH)
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
H-DL-Tle-OH
AS21853G25
AS21853G5
ACTIAS21853G25EA
0
GBP
ACTIAS21853G25
ACTIAS21853G5
DL-tert-Leucine (H-DL-Tle-OH)
DL-tert-Leucine (H-DL-Tle-OH)
Formula:
C₆H₁₃NO₂ MW: 131.2 g/mol |
MDL Number:
MFCD00065933 CAS Number: 33105-81-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...