tert-Butyl-3-aminobenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: tert-Butyl 3-aminobenzoate

AS21115G25 AS21115G5
ACTIAS21115G25EA 0 GBP
ACTIAS21115G25 ACTIAS21115G5
tert-Butyl-3-aminobenzoate
tert-Butyl 3-aminobenzoate
Formula: C₁₁H₁₅NO₂
MDL Number: MFCD00729048
CAS Number: 92146-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR