4-Bromo-1-naphthoic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS21212G1
AS21212G25
AS21212G5
ACTIAS21212G1EA
0
GBP
ACTIAS21212G1
ACTIAS21212G25
ACTIAS21212G5
4-Bromo-1-naphthoic acid
4-Bromo-1-naphthoic acid
Formula:
C₁₁H₇BrO₂ |
MDL Number:
MFCD00094221 CAS Number: 16650-55-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...