Methyl-6-bromo-2-naphthoate
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Methyl 6-bromo-2-naphthoate
AS22661G100
AS22661G25
ACTIAS22661G100EA
0
GBP
ACTIAS22661G100
ACTIAS22661G25
Methyl-6-bromo-2-naphthoate
Methyl 6-bromo-2-naphthoate
Formula:
C₁₂H₉BrO₂ MW: 265.11 g/mol |
MDL Number:
MFCD00100408 CAS Number: 33626-98-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...