tert-Butyl (diethylphosphono)acetate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22179G100 AS22179G25
ACTIAS22179G100EA 0 GBP
ACTIAS22179G100 ACTIAS22179G25
tert-Butyl (diethylphosphono)acetate
tert-Butyl (diethylphosphono)acetate
Formula: C₁₀H₂₁O₅P
MDL Number: MFCD00075414
CAS Number: 27784-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR