(3,4,5-Trimethoxyphenyl)acetonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,4,5-Trimethoxyphenylacetonitrile

AS24588G100 AS24588G25 AS24588G5
ACTIAS24588G100EA 0 GBP
ACTIAS24588G100 ACTIAS24588G25 ACTIAS24588G5
(3,4,5-Trimethoxyphenyl)acetonitrile
(3,4,5-Trimethoxyphenyl)acetonitrile
Formula: C₁₁H₁₃NO₃
MDL Number: MFCD00001912
CAS Number: 13338-63-1
EINECS: 236-388-5
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR