1-Bromo-2,5-bis(trifluoromethyl)benzene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,5-Bis(trifluoromethyl)bromobenzene, 2-Bromo-1,4-bis(trifluoromethyl)benzene

AS24894G100 AS24894G25
ACTIAS24894G100EA 82 GBP
ACTIAS24894G100 ACTIAS24894G25
1-Bromo-2,5-bis(trifluoromethyl)benzene
1-Bromo-2,5-bis(trifluoromethyl)benzene
Formula: C₈H₃BrF₆
MW: 293.01 g/mol
MDL Number: MFCD03094336
CAS Number: 7617-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR