(4-Aminophenyl)acetonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(4-Aminophenyl)acetonitrile, 4-Aminobenzyl cyanide

AS24535G100 AS24535G25
ACTIAS24535G100EA 0 GBP
ACTIAS24535G100 ACTIAS24535G25
(4-Aminophenyl)acetonitrile
(4-Aminophenyl)acetonitrile
Formula: C₈H₈N₂
MDL Number: MFCD00007912
CAS Number: 3544-25-0
EINECS: 222-587-4
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR