(2-Aminophenyl)acetonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(2-Aminophenyl)acetonitrile, 2-Aminobenzyl cyanide, (2-Aminophenyl)acetonitrile

AS23224G25 AS23224G5
ACTIAS23224G25EA 417 GBP
ACTIAS23224G25 ACTIAS23224G5
(2-Aminophenyl)acetonitrile
(2-Aminophenyl)acetonitrile
Formula: C₈H₈N₂
MW: 132.17 g/mol
MDL Number: MFCD00091082
CAS Number: 2973-50-4
EINECS: 221-015-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR