(±)-2-Amino-2-(4-bromophenyl)acetic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21498G1 AS21498G5
ACTIAS21498G1EA 41 GBP
ACTIAS21498G1 ACTIAS21498G5
(±)-2-Amino-2-(4-bromophenyl)acetic acid
(±)-2-Amino-2-(4-bromophenyl)acetic acid
Formula: C₈H₈BrNO₂
CAS Number: 129592-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR