(S)-(-)-4-Amino-2-hydroxybutyric acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(S)-4-Amino-2-hydroxybutanoic acid
AS23040G100
AS23040G25
ACTIAS23040G100EA
82
GBP
ACTIAS23040G100
ACTIAS23040G25
(S)-(-)-4-Amino-2-hydroxybutyric acid
(S)-(-)-4-Amino-2-hydroxybutyric Acid
Formula:
C₄H₉NO₃ MW: 119.12 g/mol |
MDL Number:
MFCD00674110 CAS Number: 40371-51-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...