3-Fluoro-2-nitrophenol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS31271G1
AS31271G5
ACTIAS31271G1EA
0
GBP
ACTIAS31271G1
ACTIAS31271G5
3-Fluoro-2-nitrophenol
3-Fluoro-2-nitrophenol
Formula:
C₆H₄FNO₃ MW: 157.10 g/mol |
MDL Number:
MFCD07368753 CAS Number: 385-01-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...