3-Bromo-2-chlorobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS36708G1 AS36708G5
ACTIAS36708G1EA 67 GBP
ACTIAS36708G1 ACTIAS36708G5
3-Bromo-2-chlorobenzonitrile
3-Bromo-2-chlorobenzonitrile
Formula: C₇H₃BrClN
MW: 216.4600 g/mol
MDL Number: MFCD09834776
CAS Number: 914250-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR