4-(1H-1,2,4-Triazol-1-yl)benzoic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-[1,2,4]Triazol-1-yl-benzoic acid
AS20708G1
AS20708G5
ACTIAS20708G1EA
0
GBP
ACTIAS20708G1
ACTIAS20708G5
4-(1H-1,2,4-Triazol-1-yl)benzoic acid
4-(1H-1,2,4-Triazol-1-yl)benzoic acid
Formula:
C₉H₇N₃O₂ MW: 189.1708 g/mol |
MDL Number:
MFCD02682060 CAS Number: 162848-16-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...