3-(1H-1,2,4-Triazol-1-yl)benzoic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-(1,2,4-Triazol-1-yl)benzoic Acid
AS20633G1
AS20633G5
ACTIAS20633G1EA
0
GBP
ACTIAS20633G1
ACTIAS20633G5
3-(1H-1,2,4-Triazol-1-yl)benzoic acid
3-(1H-1,2,4-Triazol-1-yl)benzoic acid
Formula:
C₉H₇N₃O₂ MW: 189.1708 g/mol |
MDL Number:
MFCD08271895 CAS Number: 167626-64-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...