4-Cyano-4-(4-fluorophenyl)cyclohexanone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(4-Fluorophenyl)-4-oxocyclohexane-1-carbonitrile

AS21022G1 AS21022G5
ACTIAS21022G1EA 131 GBP
ACTIAS21022G1 ACTIAS21022G5
4-Cyano-4-(4-fluorophenyl)cyclohexanone
4-Cyano-4-(4-fluorophenyl)cyclohexanone
Formula: C₁₃H₁₂FNO
MW: 217.25 g/mol
MDL Number: MFCD00044815
CAS Number: 56326-98-8
EINECS: 260-111-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR